Learn More
Strem, An Ascensus Company CAS# 18972-56-0. 250g. Magnesium hexafluorosilicate hexahydrate, 98%. MFCD00150250
SDP
Catalog No. 5090113959
Encompass
CAS# 18972-56-0. 250g. Magnesium hexafluorosilicate hexahydrate, 98%. MFCD00150250. Molecular Weight: 166.39 (274.48). Molecular Formula: MgSiF6∙6H2O. Color/form: white pwdr. Strem# 93-1252. http://www.strem.com/catalog/v/93-1252/
Catalog No. 50-901-13959 Supplier Strem, An Ascensus Company Supplier No. 931252250G
CAS# 18972-56-0. 250g. Magnesium hexafluorosilicate hexahydrate, 98%. MFCD00150250. Molecular Weight: 166.39 (274.48). Molecular Formula: MgSiF6∙6H2O. Color/form: white pwdr. Strem# 93-1252. http://www.strem.com/catalog/v/93-1252/