Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium hexafluorosilicate hydrate

Catalog No. p-7080484
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8901836 500 g
AA89018A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8901836 Supplier Thermo Scientific Chemicals Supplier No. 08901836
Only null left
Add to Cart

MgSiF6·xH2O

It is employed as an antiplaque agents and oral care agents. Also used for mothproofing textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula MgSiF6·xH2O
MDL Number MFCD00016196

Specifications

Melting Point 120°C (decomposition)
Physical Form Powder
Quantity 500 g
UN Number UN2853
Solubility Information Soluble in water.
Formula Weight 166.39 (Anhydrous)
Density 1.788 g/mL
Chemical Name or Material Magnesium hexafluorosilicate hydrate
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: MAGNESIUM FLUOROSILICATE
EINECSNumber 241-022-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.