Alfa Aesar™ Magnesium hexafluorosilicate hydrate

Manufacturer: Alfa Aesar™ 8901836

Quantity 500g

 View more versions of this product

Catalog No. AA8901836

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula MgSiF6·xH2O
    Formula Weight 166.39 (Anhydrous)
    MDL Number MFCD00016196
    UN Number UN2853
    Chemical Name or Material Magnesium hexafluorosilicate hydrate

    Specifications

    Formula Weight 166.39 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula MgSiF6·xH2O
    MDL Number MFCD00016196
    Quantity 500g
    Health Hazard 1 Danger
    TSCA TSCA
    Physical Form Powder
    Melting Point 120°C (dec.)
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Density 1.788g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: MAGNESIUM FLUOROSILICATE
    UN Number UN2853
    Chemical Name or Material Magnesium hexafluorosilicate hydrate

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit