Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Magnesium hydroxide | 1309-42-8 | MFCD00011104 | 99.1% | 250 G
SDP

Catalog No. 5000414480
Encompass_Preferred

Magnesium hydroxide, 99%(KT) is a biochemical agent primarily used as a non-absorbable antacid for treating acidic digestive diseases. It is supplied as a white to off-white solid.

  • Used to treat acidic digestive diseases
  • Non-absorbable antacid
  • Purity of 99.1%
  • Available in solid form
  • White to off-white appearance
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-14480 Supplier Medchemexpress LLC Supplier No. 250GHYB2218D
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Magnesium hydroxide, 99%(KT) is a biochemical agent primarily used as a non-absorbable antacid for treating acidic digestive diseases. It is supplied as a white to off-white solid.

  • Used to treat acidic digestive diseases
  • Non-absorbable antacid
  • Purity of 99.1%
  • Available in solid form
  • White to off-white appearance
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.