Learn More
Magnesium Hydroxide Suspension, 80 g/L Aqueous, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Magnesium Hydroxide |
| Color | Milky White Suspension |
| CAS | 7732-18-5, 1309-42-8 |
| CAS Max % | 92.38, 7.62 |
| Percent Purity | 77 to 83% |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | 9.5 to 10.5 |
| Quantity | 500 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.