Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Hydroxide USP/FCC Powder, Macron Fine Chemicals™

Catalog No. MK5984500 Shop All Avantor Products
Change view
Click to view available options
POLYSTORMOR; 4 x 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-5984-500 POLYSTORMOR; 4 x 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MK-5984-500 Supplier Avantor Supplier No. 598412
This item is not returnable. View return policy

Specifications

CAS 1309-42-8
Physical Form Powder
Quantity 4 x 500 g
Molecular Formula H2MgO2
MDL Number MFCD00011104
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
SMILES [OH-].[OH-].[Mg++]
IUPAC Name magnesium(2+) dihydroxide
Molecular Weight (g/mol) 58.32
PubChem CID 73981
ChEBI CHEBI:6637
Formula Weight 58.32
Grade FCC/USP
Packaging POLYSTORMOR
Chemical Name or Material Magnesium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.