Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium(II) Bis(trifluoromethanesulfonyl)imide 97.0+%, TCI America™
SDP

Catalog No. p-7133478 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28611G 1 g
M28615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28611G Supplier TCI America Supplier No. M28611G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133395-16-1
Molecular Formula C4F12MgN2O8S4
Molecular Weight (g/mol) 584.576
MDL Number MFCD14156020
InChI Key DMFBPGIDUUNBRU-UHFFFAOYSA-N
Synonym bis bis trifluoromethylsulfonyl amino magnesium, magnesium bis trifluoromethanesulfonimide, magnesium ii bis trifluoromethanesulfonyl imide, magnesium ii triflimide, magnesium bistrifluoromethanesulfonimide, magnesium bis trifluoromethylsulfonyl imide, magnesium 2+ bis bistriflylimide anion, bis trifluoromethanesulfonyl imide magnesium ii salt
PubChem CID 11851070
IUPAC Name magnesium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Mg+2]

Specifications

Color White
Quantity 1 g
UN Number 1759
Formula Weight 584.58
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Magnesium(II) Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.