Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Iodide, MP Biomedicals

Catalog No. ICN202698 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202698 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202698 Supplier MP Biomedicals, Inc Supplier No. 0520269825
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent for the production of olefins from cyclic sulfates of vic-diols.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-31-0
Molecular Formula I2Mg
Molecular Weight (g/mol) 278.114
InChI Key BLQJIBCZHWBKSL-UHFFFAOYSA-L
Synonym magnesium iodide mgi2, mgi2, diiodomagnesium, magnesium iodide, magnesium iodide, powder trace metals basis, magnesium iodide, anhydrous, beads,-10 mesh trace metals basis, magnesium iodide, ultra dry metals basis , ampouled under argon, magnesium iodide, ultra dry metals basis ,-10 mesh beads, ampouled under argon
PubChem CID 66322
IUPAC Name magnesium;diiodide
SMILES [Mg+2].[I-].[I-]

Specifications

Physical Form Powder
Quantity 25 g
Formula Weight 278.12
Chemical Name or Material Magnesium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.