Learn More
Medchemexpress LLC Magnesium isoglycyrrhizinate hydrate | 658701-67-8 | C42H68MgO20 | 5 MG
SDP
Catalog No. 5000376928
Encompass_Preferred
Magnesium isoglycyrrhizinate hydrate is the active component found in licorice plant (Glycyrrhiza glabra). It exhibits anti-inflammatory activity through the NLRP3 inflammasome pathway and attenuates chronic obstructive pulmonary disease in rats. This product is for research use only.
  • Active component of licorice plant (Glycyrrhiza glabra)
  • Exhibits anti-inflammatory activity
  • Acts through the NLRP3 inflammasome pathway
  • Attenuates chronic obstructive pulmonary disease in rats
Catalog No. 50-003-76928 Supplier Medchemexpress LLC Supplier No. HY1547955MG
Magnesium isoglycyrrhizinate hydrate is the active component found in licorice plant (Glycyrrhiza glabra). It exhibits anti-inflammatory activity through the NLRP3 inflammasome pathway and attenuates chronic obstructive pulmonary disease in rats. This product is for research use only.
  • Active component of licorice plant (Glycyrrhiza glabra)
  • Exhibits anti-inflammatory activity
  • Acts through the NLRP3 inflammasome pathway
  • Attenuates chronic obstructive pulmonary disease in rats