Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Magnesium lactate | 18917-93-6 | MFCD00050751 | 98.0% | 200.43 g/mol | C6H8MgO6 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1350555MG

Encompass_Preferred

Magnesium lactate is a water-soluble magnesium salt used as a magnesium source in formulation and supplementation research. It is commonly applied in food, beverage, and pharmaceutical research settings where a soluble, bioavailable magnesium compound is required. The material is provided in small laboratory pack sizes and is intended for research use only.

  • Purity: 98.0%.
  • Molecular formula: C6H8MgO6.
  • Molecular weight: 200.43 g/mol.
  • CAS number: 18917-93-6.
  • Suitable for formulation and analytical method development.
  • Supplied in small milligram pack sizes for laboratory use.

Catalog No. 50-002-22482


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.