Learn More
Selleck Chemical LLC Magnesium lactate trihydrate S4829-1g
SDP
Catalog No. 503110695
Encompass_Preferred
Magnesium lactate the magnesium salt of lactic acid is a mineral supplement that is added to some food and beverages as an acidity regulator
Catalog No. 50-311-0695 Supplier Selleck Chemical LLC Supplier No. S48291G
Magnesium lactate the magnesium salt of lactic acid is a mineral supplement that is added to some food and beverages as an acidity regulator