Learn More
Medchemexpress LLC Magnesium Lithospermate B | 122021-74-3 | 99.49% | 740.90 | 1 ML
SDP
Catalog No. 5000435825
Encompass_Preferred
Magnesium Lithospermate B is a derivative of caffeic acid tetramer and an inhibitor of Na+/K+ ATPase, extracted from *Salviae miltiorrhizae*. It is used for research on cardiovascular diseases, protecting against glucose-induced intracellular oxidative damage, and suppressing neuroinflammation and neurodegeneration.
  • Used in research for cardiovascular diseases
  • Protects against glucose-induced intracellular oxidative damage
  • Suppresses neuroinflammation and attenuates neurodegeneration
  • Decreases LDH activity and increases SOD activity in vitro
  • Enhances proliferation and promotes differentiation of neural stem cells
  • Reduces renal damage from oxidative stress in vivo
  • Promotes neurogenesis and improves memory in PD models
Catalog No. 50-004-35825 Supplier Medchemexpress LLC Supplier No. HY12641510MM/1ML
Magnesium Lithospermate B is a derivative of caffeic acid tetramer and an inhibitor of Na+/K+ ATPase, extracted from *Salviae miltiorrhizae*. It is used for research on cardiovascular diseases, protecting against glucose-induced intracellular oxidative damage, and suppressing neuroinflammation and neurodegeneration.
  • Used in research for cardiovascular diseases
  • Protects against glucose-induced intracellular oxidative damage
  • Suppresses neuroinflammation and attenuates neurodegeneration
  • Decreases LDH activity and increases SOD activity in vitro
  • Enhances proliferation and promotes differentiation of neural stem cells
  • Reduces renal damage from oxidative stress in vivo
  • Promotes neurogenesis and improves memory in PD models