Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium matrix modifier solution, 99.9995%, Solstice

Catalog No. 60048476 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-476 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-476 Supplier Solstice Supplier No. 428892100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TraceSELECT™, 2% Mg in <5 wt. % HNO3, 99.9995%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-18-9
Molecular Weight (g/mol) 256.41 g/moL
MDL Number MFCD00085308
InChI Key FYYHWMGAXLPEAU-UHFFFAOYSA-N
Synonym Magnesium Nitrate, Nitric Acid Solution, Magnesium Modifier
IUPAC Name Magnesium Nitrate, Nitric Acid Solution, Magnesium Modifier

Specifications

Quantity 100 mL
Assay Percent Range 99.9995%
UN Number UN3264
Beilstein 4948473
InChI Formula 1S/Mg
Percent Purity 100.00%
Density 1.093g/mL at 25°C
Chemical Name or Material Magnesium matrix modifier solution
Hazard Statement P220-P305 + P351 + P338
DOTInformation 8/PGIII
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.