Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Metal (Ribbon/Turnings)
SCI_ED

Catalog No. S72223D Shop All Fisherbrand Products
Change view
Click to view available options
Ribbon; 2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S72223D Ribbon; 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S72223D

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Gray
CAS 7439-95-4
Quantity 2.5 g
Form Ribbon/Turnings
Molecular Formula Mg
MDL Number MFCD00085308
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES [Mg]
IUPAC Name magnesium
Molecular Weight (g/mol) 24.30
PubChem CID 5462224
ChEBI CHEBI:25107
Formula Weight 24.3
Grade Laboratory
Chemical Name or Material Magnesium Metal (Ribbon/Turnings)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.