Learn More
Strem, An Ascensus Company CAS# 13767-03-8. 50g. Magnesium molybdate (99.9%-Mo). MFCD00016195
SDP
Catalog No. 5090113946
Encompass
CAS# 13767-03-8. 50g. Magnesium molybdate (99.9%-Mo). MFCD00016195. Molecular Weight: 184.25. Molecular Formula: MgMoO4. Color/form: white pwdr. Strem# 93-1235. http://www.strem.com/catalog/v/93-1235/
Catalog No. 50-901-13946 Supplier Strem, An Ascensus Company Supplier No. 93123550G
CAS# 13767-03-8. 50g. Magnesium molybdate (99.9%-Mo). MFCD00016195. Molecular Weight: 184.25. Molecular Formula: MgMoO4. Color/form: white pwdr. Strem# 93-1235. http://www.strem.com/catalog/v/93-1235/