Learn More
Strem, An Ascensus Company CAS# 13446-18-9. 250g. Magnesium nitrate hexahydrate, 99% (ACS). MFCD00149779
SDP
Catalog No. 5090113951
Encompass
CAS# 13446-18-9. 250g. Magnesium nitrate hexahydrate, 99% (ACS). MFCD00149779. Molecular Weight: 148.32 (256.41). Molecular Formula: Mg(NO3)2∙6H2O. Color/form: white xtl. Strem# 93-1238. http://www.strem.com/catalog/v/93-1238/
Catalog No. 50-901-13951 Supplier Strem, An Ascensus Company Supplier No. 931238250G
CAS# 13446-18-9. 250g. Magnesium nitrate hexahydrate, 99% (ACS). MFCD00149779. Molecular Weight: 148.32 (256.41). Molecular Formula: Mg(NO3)2∙6H2O. Color/form: white xtl. Strem# 93-1238. http://www.strem.com/catalog/v/93-1238/