Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Nitrate, Hexahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18611435 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-435 500 g Amber Glass Bottle
18-611-434 125 g Amber Glass Bottle
18-611-436 2.5 kg Poly Pail
18-611-437 12 kg Poly Pail
18-611-438 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-611-435 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1050500GM
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
IUPAC Name magnesium(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

CAS Max % 1
pH 5.0 to 8.2
Quantity 500 g
Percent Purity 98 to 102%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.