Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium oxalate dihydrate, Thermo Scientific™, 99.999% (metals basis)

Catalog No. AA1299304
Change view
Click to view available options
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA1299304 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1299304 Supplier Thermo Scientific Chemicals Supplier No. 1299304

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 547-66-0
Quantity 2g
Molecular Formula C2H2MgO4+2
MDL Number MFCD00287279
UN Number UN3288
Merck Index 14,5676
Synonym magnesium oxalate, magnesium oxalate 1:1, magnesium oxalate mgc2o4, oxalic acid, magnesium salt 1:1, ethanedioic acid, magnesium salt 1:1
Solubility Information Soluble in dilute mineral acid; Practically insoluble in water; Insoluble in alcohol
InChI Key UHNWOJJPXCYKCG-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O.[Mg+2]
IUPAC Name magnesium;oxalic acid
Molecular Weight (g/mol) 114.339
PubChem CID 54611841
Formula Weight 148.36 (112.32 Anhydrous)
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Assay (metals basis)
Chemical Name or Material Magnesium oxalate dihydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.