Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Oxalate, MP Biomedicals

Catalog No. ICN202908 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202908 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202908 Supplier MP Biomedicals, Inc Supplier No. 0520290880

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 547-66-0
Molecular Formula C2H2MgO4+2
Molecular Weight (g/mol) 114.339
InChI Key UHNWOJJPXCYKCG-UHFFFAOYSA-N
Synonym magnesium oxalate, magnesium oxalate 1:1, magnesium oxalate mgc2o4, oxalic acid, magnesium salt 1:1, ethanedioic acid, magnesium salt 1:1
PubChem CID 54611841
IUPAC Name magnesium;oxalic acid
SMILES C(=O)(C(=O)O)O.[Mg+2]

Specifications

Quantity 100 g
Formula Weight 148.36
Chemical Name or Material Magnesium Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.