Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium oxide, 2 part ceramic adhesive paste

Catalog No. AA43135A4
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43135A4 2.5 kg
AA4313536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43135A4 Supplier Thermo Scientific Chemicals Supplier No. 043135A4
Add to Cart

High Temperature Adhesive For Bonding Ceramic To Metal

High temperature adhesive for bonding ceramic to metal

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Mix ratio (w/w): 1.5:1 powder to liquid. Cure conditions: 1-4 hours at room temperature, 2 hours at 94°C. Components: Magnesium oxide, 2 part ceramic adhesive paste (L) and Magnesium oxide, 2 part ceramic adhesive paste (P)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium oxide
Name Note Mix ratio (w/w): 1.5:1 powder to liquid. Cure conditions: 1-4 hours at room temperature, 2 hours at 94°C
CAS 1309-48-4
Quantity 2.5 kg
Molecular Formula MgO
MDL Number MFCD00011109
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES O=[Mg]
IUPAC Name oxomagnesium
Molecular Weight (g/mol) 40.30
PubChem CID 14792
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.