Learn More
Magnesium oxide, 96%, light, Thermo Scientific Chemicals
Description
Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, a protective coating in plasma displays, an oxide barrier in spin-tunneling devices, an optical material, a component of tubular construction in heating elements, and also in libraries for preserving books. Magnesium oxide microspheres used in enrichment method of polycyclic aromatic hydrocarbons.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 3600°C |
Melting Point | 2852°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 96% |
Solubility Information | Soluble in acid and ammonia. Insoluble in water and ethanol. |
Formula Weight | 40.3 |
Refractive Index | 1.736 |
Appearance | White powder |
Show More |
Safety and Handling
EINECSNumber | 215-171-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.