Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium oxide, 99.99%, (trace metal basis), extra pure, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1309-48-4
MgO
40.29
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide,magnesia,seawater magnesia,causmag,granmag,maglite,periclase,seasorb,animag
14792
oxomagnesium
O=[Mg]
1309-48-4
MgO
40.29
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide,magnesia,seawater magnesia,causmag,granmag,maglite,periclase,seasorb,animag
14792
oxomagnesium
O=[Mg]
For ultra trace analysis
Assay Percent Range 99.99%
Linear Formula MgO
Fieser 01,633; 02,256; 07,221; 16,200
Merck Index 15, 5741
Solubility Information Solubility in water: 6.2mg/l (20°C),reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids,insoluble in alcohol
Formula Weight 40.29
Physical Form Powder
Percent Purity 99.99%
Grade Trace Metal Basis
Density 3.5800g/mL
Packaging Glass bottle
Solubility Solubility in water: 6.2 mg/l (20°C), reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids, insoluble in alcohol
Specific Gravity 3.58
Total Trace Metal Impurities 0.01% max
Boiling Point 3600.0°C
Color White
Melting Point 2800.0°C
Quantity 50g
Chemical Name or Material Magnesium oxide, Extra Pure
Show More Show Less

EINECSNumber : 215-171-9