Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Oxide, ACS Grade, For Analysis (max.0.001% SO4) EMSURE™, MilliporeSigma™

Catalog No. M1058660100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1058660100 100 g Plastic bottle
M1058660500 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1058660100 Supplier MilliporeSigma Supplier No. 1.05866.0100
Add to Cart
Add to Cart
For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Physical Form Solid
Quantity 100 g
Formula Weight 40.3
Grade ACS
Odor Odorless
Packaging Plastic bottle
Appearance White solid
Chemical Name or Material Magnesium oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.