Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Oxide Heavy Powder USP, DPL

Catalog No. 502985593
Encompass
Catalog No. 50-298-5593 Supplier Thermo Scientific Production Chemicals and Services Supplier No. D503046001SPH
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1309-48-4
Molecular Weight (g/mol) 40.3 g/mol

Specifications

Physical Form Powder
Quantity 25 kg
Percent Purity 96.0 to 100.5%
Grade USP
Packaging Polylined paperbag
Chemical Name or Material Magnesium oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.