Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Oxide, Powder, Technical, Spectrum™ Chemical
SDP

Catalog No. 18611443 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 lb.
45 kg
Packaging:
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-443 2.5 kg Fiber Drum
18-611-441 500 g Fiber Drum
18-611-442 50 lb. Fiber Drum
18-611-439 12 kg Fiber Drum
18-611-440 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-611-443 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M10572.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1309-48-4

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 2.5 kg
Grade Technical
Packaging Fiber Drum
Chemical Name or Material Magnesium Oxide, Powder

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.