Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium perchlorate ACS100G
Catalog No. 502898659
Encompass
Magnesium perchlorate (Mg(ClO4)2) is widely used as drying agent for gases. It can remove water from gases (with no organic contaminants) at the rate of 0.001mgwater/l.
Catalog No. 50-289-8659 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 222283100G
Magnesium perchlorate (Mg(ClO4)2) is widely used as drying agent for gases. It can remove water from gases (with no organic contaminants) at the rate of 0.001mgwater/l.