Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium perchlorate ACS500G
Catalog No. 502899485
Encompass
Magnesium perchlorate (Mg(ClO4)2) is widely used as drying agent for gases. It can remove water from gases (with no organic contaminants) at the rate of 0.001mgwater/l.
Catalog No. 50-289-9485 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 222283500G
Magnesium perchlorate (Mg(ClO4)2) is widely used as drying agent for gases. It can remove water from gases (with no organic contaminants) at the rate of 0.001mgwater/l.