Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Phenoxyacetate Dihydrate 98.0+%, TCI America™
SDP

Catalog No. P010825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P010825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P010825G Supplier TCI America Supplier No. P010825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 76172-73-1
Molecular Formula C16H20MgO8
Molecular Weight (g/mol) 364.633
MDL Number MFCD00059853
InChI Key ZFPUMEQWQQBGMQ-UHFFFAOYSA-N
Synonym Phenoxyacetic Acid Magnesium Salt
PubChem CID 131884797
IUPAC Name magnesium;2-phenoxyacetic acid;dihydrate
SMILES C1=CC=C(C=C1)OCC(=O)O.C1=CC=C(C=C1)OCC(=O)O.O.O.[Mg]

Specifications

Color White
Quantity 25 g
Formula Weight 326.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Magnesium Phenoxyacetate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.