Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Reagent, LabChem™

Catalog No. LC164707 Shop All LabChem Products
Change view
Click to view available options
Quantity:
125 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC164707 125 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC164707 Supplier LabChem, Inc. Supplier No. LC164707
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Magnesium Reagent
Color Purple-Red
CAS 7732-18-5, 1310-73-2, 74-39-5
CAS Max % 98.5, 1, 0.5
Physical Form Liquid
Quantity 125 mL
Linear Formula C12H9N3O4
UN Number UN1824
Solubility Information Soluble in water
Formula Weight 259.22
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.