Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Reagent, TAPPI for Mineral Filler, Ricca Chemical
SDP

Catalog No. R4487000120 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4487000120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4487000120 Supplier Ricca Chemical Company Supplier No. R4487000120A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2 , 74-39-5
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Red
CAS Min % 97.0, 0.97, 0.48
CAS Max % 100.0, 1.01, 0.5
Physical Form Liquid
Quantity 120 mL
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
Chemical Name or Material Magnesium Reagent
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.