Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Salicylate, Powder, USP, 98-103%, Spectrum™ Chemical
SDP

Catalog No. 18604107 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
75 g
125 g
500 g
2.5 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-107 75 g Poly Bottle
18-604-103 125 g Poly Bottle
18-604-104 500 g Poly Bottle
18-604-105 2.5 kg Poly Bottle
18-604-106 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-107 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MA12875GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MA128, 18917-95-8, C14H10MgO6·4H2O

Spectrum™ Chemical Magnesium Salicylate, Powder, USP is a non-steroidal anti-inflammatory drug (NSAID) in a group of drugs called salicylates. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 18917-95-8
Molecular Formula C14H18MgO10
Molecular Weight (g/mol) 370.59
MDL Number MFCD01766176,MFCD00045815
InChI Key NBQBEWAYWAMLJJ-UHFFFAOYSA-L
IUPAC Name magnesium(2+) bis(2-hydroxybenzoate) tetrahydrate
SMILES O.O.O.O.[Mg++].OC1=CC=CC=C1C([O-])=O.OC1=CC=CC=C1C([O-])=O

Specifications

Physical Form Crystalline Powder or Needles
CAS Max % 100%
Quantity 75 g
Packaging Poly Bottle
Percent Purity 98 to 103%
Grade USP
Chemical Name or Material Magnesium Salicylate, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.