Learn More
Sigma Organic Chemistry Magnesium salicylate te | 1G | 18917-95-8 | MFCD01766176
Supplier: Sigma Organic Chemistry CDS0012381G
Magnesium salicylate te , 1G
About this Item:
CAS #: 18917-95-8
MDL #: MFCD01766176
Molecular Weight: 370.59
UNSPSC Code: 12352200
Chemical Formula: C14H18MgO10
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.