Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium silicate, 99% (metals basis)

Catalog No. p-4407621
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4380718 50 g
AA4380730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4380718 Supplier Thermo Scientific Chemicals Supplier No. 04380718
Only null left
Add to Cart

CAS: 1343-88-0 | MgO3Si | 100.39 g/mol

Magnesium silicate is employed for polyether adsorbent, which has applications of adsorption and decoloring, adsorbent in gas chromatography, in vitamin analysis, antibiotic processing, ant caking agent, used in cosmetics as talcum & baby powder, general modified plastics, mid-range insulation materials, EVA shoes material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
MDL Number MFCD00078249
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Specifications

Melting Point 1910°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99% (Metal basis)
Solubility Information Insoluble in water
Formula Weight 140.71
Odor Odorless
Appearance White powder
Density 3.21 g/cm3 at 20°C
Chemical Name or Material Magnesium silicate
Show More Show Less
Hazard Category H332: Harmful if inhaled.
EINECSNumber 215-681-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.