Learn More
SPEX CERTIPREP LLC 3P MAGNESIUM STD 100PPM/100ML
Catalog No. NC9799484
Encompass_Preferred
(HPS by Zeptometrix) 100 ug/mL Magnesium in H2O, Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9799484 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICMG100
(HPS by Zeptometrix) 100 ug/mL Magnesium in H2O, Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.