Learn More
Medchemexpress LLC Magnesium stearate | 557-04-0 | MFCD00036391 | 99.0% | 591.24 | C36H70MgO4 | 500g
SDP
Catalog No. 5000268134
Encompass_Preferred
Magnesium stearate is a commonly used pharmaceutical lubricant But Magnesium stearate leads to an adverse effect on bonding between particles Magnesium stearate can be used as an excipient such as lubricant Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1][2]
Catalog No. 50-002-68134 Supplier Medchemexpress LLC Supplier No. HYY1054500G
Magnesium stearate is a commonly used pharmaceutical lubricant But Magnesium stearate leads to an adverse effect on bonding between particles Magnesium stearate can be used as an excipient such as lubricant Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1][2]