Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Stearate, NF, BP, JP, 4-5%, Spectrum™ Chemical
SDP

Catalog No. 18604113 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-113 500 g Poly Bottle
18-604-115 2.5 kg Poly Pail
18-604-114 6 x 500 g Poly Bottle
18-604-116 4 x 2.5 kg Poly Pail
18-604-117 12 kg Fiber Drum
18-604-118 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-113 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MA130500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

MA130, 557-04-0

Spectrum™ Chemical Magnesium Stearate, NF, BP, JP is used as an anti-adherent in the manufacture of of medical capsules, tablets, and powders. Spectrum Chemical manufactured NF, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 557-04-0
Molecular Formula C36H70MgO4
Molecular Weight (g/mol) 591.26
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
IUPAC Name magnesium(2+) dioctadecanoate
SMILES [Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 4 to 5%
Grade Multi-Compendial
Loss on Drying 6%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.