Learn More
Magnesium Stearate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Catalog No. 111004770 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2.5 g
Catalog No. Quantity
11-100-4770 2.5 g
1 options
Catalog No. 11-100-4770 Supplier MilliporeSigma Supelco Supplier No. PHR13632.5G
Add to Cart
Add to Cart
Magnesium stearate is commonly utilized as a lubricant in pharmaceutical formulations to avoid the excipient blend from the formulation contents from sticking to the manufacturing equipment.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Type Certified Reference Material
Quantity 2.5 g
Grade Certified Reference Material