Learn More
SPEX CERTIPREP LLC 3P MAGNESIUM STNDRD 250ML 1000PPM
Catalog No. NC9203932
Encompass_Preferred
(HPS by Zeptometrix) Magnesium standard; 1000ug/mL; 250 mL, Matrix: 2% HNO3, High Purity Standards
Catalog No. NC9203932 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000311250
(HPS by Zeptometrix) Magnesium standard; 1000ug/mL; 250 mL, Matrix: 2% HNO3, High Purity Standards