Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate, 2.25% (w/v), Ricca Chemical
SDP

Catalog No. 453032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4530-32 1 L Natural Poly Bottle
4530-16 500 mL Natural Poly Bottle
R45300004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 4530-32 Supplier Ricca Chemical Company Supplier No. 453032
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10034-99-8 , 7732-18-5
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Color Colorless
Percent Purity 2.25%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 2.25% (w/v)
Chemical Name or Material Magnesium Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.