Learn More
Medchemexpress LLC Magnesium sulfate | 7487-88-9 | MFCD00011110 | 99.6% | 50g
SDP
Catalog No. 5000267053
Encompass_Preferred
Magnesium sulfate is a calcium antagonist and a potent L-type calcium channel inhibitor as well as a tocolytic Magnesium sulfate has anti-inflammatory anticonvulsant vasodilatory and neuroprotective effects Magnesium sulfate can be used in the research of diseases such as preeclampsia/eclampsia[1][2][3][4][5][6]
Catalog No. 50-002-67053 Supplier Medchemexpress LLC Supplier No. HYY126750G
Magnesium sulfate is a calcium antagonist and a potent L-type calcium channel inhibitor as well as a tocolytic Magnesium sulfate has anti-inflammatory anticonvulsant vasodilatory and neuroprotective effects Magnesium sulfate can be used in the research of diseases such as preeclampsia/eclampsia[1][2][3][4][5][6]