Learn More
Strem, An Ascensus Company CAS# 7487-88-9. 1kg. Magnesium sulfate, anhydrous, 99+%. MFCD00011110
SDP
Catalog No. 5090113962
Encompass
CAS# 7487-88-9. 1kg. Magnesium sulfate, anhydrous, 99+%. MFCD00011110. Molecular Weight: 120.39. Molecular Formula: MgSO4. Color/form: white pwdr. Strem# 93-1255. http://www.strem.com/catalog/v/93-1255/
Catalog No. 50-901-13962 Supplier Strem, An Ascensus Company Supplier No. 9312551000G
CAS# 7487-88-9. 1kg. Magnesium sulfate, anhydrous, 99+%. MFCD00011110. Molecular Weight: 120.39. Molecular Formula: MgSO4. Color/form: white pwdr. Strem# 93-1255. http://www.strem.com/catalog/v/93-1255/