Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium sulfate anhydrou
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M75062KG
Encompass
Magnesium sulfate is a magnesium salt with tocolytic ability. Its role to induce an analgesic effect during intraoperative and postoperative periods has been investigated. It is reported to prevent eclampsia. Its ability to remove water from water-miscible solvents by salting-out phase separation has been studied.
Catalog No. 50-290-1995