Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium sulfate anhydrou
Catalog No. 502902153
Encompass
Magnesium sulfate is a magnesium salt with tocolytic ability. Its role to induce an analgesic effect during intraoperative and postoperative periods has been investigated. It is reported to prevent eclampsia. Its ability to remove water from water-miscible solvents by salting-out phase separation has been studied.
Catalog No. 50-290-2153 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M750612KG
Magnesium sulfate is a magnesium salt with tocolytic ability. Its role to induce an analgesic effect during intraoperative and postoperative periods has been investigated. It is reported to prevent eclampsia. Its ability to remove water from water-miscible solvents by salting-out phase separation has been studied.