Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium sulfate anhydrou1KG
Catalog No. 502898844
Encompass
Magnesium sulfate is a magnesium salt with tocolytic ability. Its role to induce an analgesic effect during intraoperative and postoperative periods has been investigated. It is reported to prevent eclampsia. Its ability to remove water from water-miscible solvents by salting-out phase separation has been studied.
Catalog No. 50-289-8844 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M75061KG
Magnesium sulfate is a magnesium salt with tocolytic ability. Its role to induce an analgesic effect during intraoperative and postoperative periods has been investigated. It is reported to prevent eclampsia. Its ability to remove water from water-miscible solvents by salting-out phase separation has been studied.