Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium sulfate anhydrou25KG
Catalog No. 502914199
Encompass
Magnesium sulfate is a magnesium salt with tocolytic ability. Its role to induce an analgesic effect during intraoperative and postoperative periods has been investigated. It is reported to prevent eclampsia. Its ability to remove water from water-miscible solvents by salting-out phase separation has been studied.
Catalog No. 50-291-4199 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M750625KG
Magnesium sulfate is a magnesium salt with tocolytic ability. Its role to induce an analgesic effect during intraoperative and postoperative periods has been investigated. It is reported to prevent eclampsia. Its ability to remove water from water-miscible solvents by salting-out phase separation has been studied.