Learn More
Alkali Scientific Magnesium Sulfate Anhydrous, 25 Kg
SDP
Catalog No. 5000659824
Encompass_Preferred
Magnesium Sulfate Anhydrous is a highly pure, granular reagent that is widely used in laboratory experiments and chemical synthesis. In its anhydrous form, this compound is an essential component in a variety of applications, including pharmaceutical formulations, chemical reactions, and laboratory assays. It is primarily used to provide magnesium ions in solutions and as a desiccant due to its ability to absorb water. This 25 kg quantity is suitable for bulk laboratory use where consistent performance and high purity are required. Magnesium Sulfate Anhydrous is particularly important in reactions that require a stable, low-humidity environment, and it is commonly used in preparation for various chemical and biochemical processes.
Catalog No. 50-006-59824 Supplier Alkali Scientific Supplier No. ESI001CP
Magnesium Sulfate Anhydrous is a highly pure, granular reagent that is widely used in laboratory experiments and chemical synthesis. In its anhydrous form, this compound is an essential component in a variety of applications, including pharmaceutical formulations, chemical reactions, and laboratory assays. It is primarily used to provide magnesium ions in solutions and as a desiccant due to its ability to absorb water. This 25 kg quantity is suitable for bulk laboratory use where consistent performance and high purity are required. Magnesium Sulfate Anhydrous is particularly important in reactions that require a stable, low-humidity environment, and it is commonly used in preparation for various chemical and biochemical processes.