Learn More
Alkali Scientific Magnesium Sulfate Anhydrous, 5 Kg
SDP
Catalog No. 5000661474
Encompass_Preferred
Magnesium Sulfate Anhydrous is a highly concentrated, dry form of magnesium sulfate, providing a pure source of magnesium ions for laboratory and industrial applications. This 5 kg packaging is ideal for bulk use in research labs that need magnesium sulfate for a variety of processes. Commonly used as a drying agent, a desiccant, and in the preparation of various chemical reactions, this compound is crucial in pharmaceutical, biochemical, and environmental analysis. Magnesium Sulfate Anhydrous is also employed in agricultural applications and as a supplement in nutrient solutions, as well as for preparing solutions in laboratory experiments where high purity is required.
Catalog No. 50-006-61474 Supplier Alkali Scientific Supplier No. NES003CP
Magnesium Sulfate Anhydrous is a highly concentrated, dry form of magnesium sulfate, providing a pure source of magnesium ions for laboratory and industrial applications. This 5 kg packaging is ideal for bulk use in research labs that need magnesium sulfate for a variety of processes. Commonly used as a drying agent, a desiccant, and in the preparation of various chemical reactions, this compound is crucial in pharmaceutical, biochemical, and environmental analysis. Magnesium Sulfate Anhydrous is also employed in agricultural applications and as a supplement in nutrient solutions, as well as for preparing solutions in laboratory experiments where high purity is required.