Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Magnesium Sulfate Anhydrous
SCI_ED

Catalog No. S25414B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25414B 500 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25414B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity magnesium sulfate anhydrous ideal for educational labs and classroom experiments.

  • Educational Grade: Designed for instructional use in science classrooms and teaching laboratories
  • Chemical Name: Magnesium Sulfate, Anhydrous Formula: MgSO4
  • Formula Weight: 120.37
  • Form: Solid powder
  • Density: 2.65 g/mL
  • Quantity: 500 grams
  • Packaging: Secure solid-form container
  • Note: For educational use only, Certificate of Analysis (C of A) not available

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
IUPAC Name magnesium(2+) sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Physical Form Powder, Crystals and/or Chunks
Quantity 500 g
Linear Formula MgSO4
Formula Weight 120.37
Packaging Solid
Density 2.65g/mL
Chemical Name or Material Magnesium Sulfate, Anhydrous
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.