Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate AR Powder, Macron Fine Chemicals™

Catalog No. MK6070500 Shop All Avantor Products
Change view
Click to view available options
POLYSTORMOR; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-6070-500 POLYSTORMOR; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MK-6070-500 Supplier Avantor Supplier No. 607012

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7487-88-9
Physical Form Powder
Quantity 500 g
Molecular Formula MgO4S
MDL Number MFCD00011110
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES [Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium(2+) sulfate
Molecular Weight (g/mol) 120.36
PubChem CID 24083
ChEBI CHEBI:32599
Formula Weight 120.37
Packaging POLYSTORMOR
Chemical Name or Material Magnesium Sulfate, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.