Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate Dried, MACCO

Catalog No. 502985596
Encompass
Change view
Click to view available options
Quantity:
12 kg
45 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-298-5596 45 kg
50-298-5651 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-298-5596 Supplier Thermo Scientific Production Chemicals and Services Supplier No. M51204000450
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22189-08-8
Molecular Weight (g/mol) 138.38 g/mol

Specifications

Physical Form Solid
Quantity 45 kg
Percent Purity 62 to 78%
Grade USP
Packaging 45 kg poly drum with 2xPE liner
Chemical Name or Material Magnesium sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.